Learn More
Andwin ScientificSupplier Diversity Partner TIP PK/RK E GR10UL PK1152 DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 53503816
The 0.1–10 µL Ultra-Micro Pipet Tips are made to be use with standard size micropipettors. These tips are free from RNase and DNase, are nonsterile, and are autoclavable. This product is sold in a pack of 1152 tips.
Catalog No. 50-112-6047