Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIPIN Mouse anti-Human, Clone: OTI2H6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526571

Catalog No. PIMA526571


Add to cart

Description

Description

Required for normal progression of S-phase. Important for cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.
Specifications

Specifications

TIPIN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BVW5
TIMELESS interacting protein, FLJ20516
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54962
Immunohistochemistry (Paraffin), Western Blot
OTI2H6
Unconjugated
TIPIN
Liquid
TIPIN
Full length human recombit protein of TIPIN produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit