Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Tiplaxtinin
Plasminogen activator inhibitor-1 (PAI-1) inhibitor
$200.68 - $841.00
Specifications
Chemical Name or Material | α-Oxo-1-(phenylmethyl)-5-[4-(trifluoromethoxy)phenyl]-1H-indole-3-acetic acid |
---|---|
CAS | 393105-53-8 |
Synonym | PAI 039, tiplasinin |
Target | Additional Synthetic Protease Inhibitors |
Molecular Formula | C24H16F3NO4 |
Description
Plasminogen activator inhibitor-1 (PAI-1) inhibitor (IC50 = 2.7 μM). Induces vascular smooth muscle cell apoptosis in vitro and reduces carotid artery neointimal formation in a rat vascular injury model. Promotes thrombolysis in rat and dog models of acute arterial thrombosis. Orally available.Specifications
α-Oxo-1-(phenylmethyl)-5-[4-(trifluoromethoxy)phenyl]-1H-indole-3-acetic acid | |
PAI 039, tiplasinin | |
C24H16F3NO4 | |
>98% |
393105-53-8 | |
Additional Synthetic Protease Inhibitors | |
439.38 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title