Learn More
Tecan 200L MCA96 Tip NF Nest
Supplier: Tecan 30038619
Genuine Tecan 200 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, nested 8 stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 5 tip boxes x 96 tips/rack, 3,840 tips/sales unit. Nested disposable tips offer improved throughput and walk way time due to nested format allowing more tips per carrier. They also give you increased flexibility on the work deck due to ANSI format and are more environmentally friendly, through significant weigh and waste reduction. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.