Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB I, for Fluoride ISE, Reagents

Catalog No. 01336224
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-224 20 L
01-336-225 1 L
01-336-223 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-224 Supplier Reagents Holdings Llc Supplier No. C734020020F
Only null left
Add to Cart
Add to Cart

TISAB I, for Fluoride ISE, Reagent, use for Fluoride analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Total Ionic Strength Adjustment Buffer
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Application) Fluoride Analysis
For Use With (Equipment) Fluoride Meter
pH 5 to 5.5
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.1
Sterility Non-sterile
Grade Reagent
Chemical Name or Material TISAB I, for Fluoride ISE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.