Learn More
TISAB IV solution, For determination of fluoride, MilliporeSigma™ Supelco™
Total ionic strength adjustment buffers (TISAB), is generally used for the microdetermination of fluorides.
Manufacturer:  MilliporeSigma Supelco 345744X2.5L
 View more versions of this product
Catalog No. 11-100-2583
Add to cart
Description
Description
Specifications
Specifications
64-19-7
4 x 2.5 L
For determination of fluoride