Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Tissue Forceps, Sklar 16 in., 2x3 Teeth, Straight, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 610117
Forceps, Tissue Forceps, Sklar, 16 in., 2x3 Teeth, Straight, Stainless Steel, EA. Sklar Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference. This product is straight with 2x3 teeth, and a length of 16 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-8850