Learn More
Thermo Scientific™ Tissue-Loc™ HistoScreen™ Cassettes Available on GSA/VA Contract for Federal Government customers only.
Handle small biopsies and fragile specimens with Thermo Scientific™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
Manufacturer:  Thermo Scientific™ C1000TN
 View more versions of this product
Catalog No. 23-900-653
  Check Availability
Add to cart
Specifications
Description
Handle small biopsies and fragile specimens with Thermo Scientific™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
Description & Specifications
Specifications
Tan