Learn More
Thermo Scientific™ Tissue-Loc™ HistoScreen™ Cassettes Available on GSA/VA Contract for Federal Government customers only.
Handle small biopsies and fragile specimens with Thermo Scientific™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
$424.00 - $1065.00
Specifications
Type HistoScreen™ Cassettes, Mesh Pore Insert
View More Specs
Products 10
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-045348
Accessories View Documents
Thermo Scientific™
C1000BL
Blue
Pack of 250 for $429.50
Case of 4 PK for $1,509.00
Add to cart
22-045347
Accessories View Documents
Thermo Scientific™
C1000GR
Green
Pack of 250 for $429.50
Case of 4 PK for $1,502.00
Add to cart
22-045349
Accessories View Documents
Thermo Scientific™
C1000GY
Gray
Pack of 250 for $424.00
Case of 4 PK for $1,465.15
Add to cart
13-874-609 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
C1000LI
Lilac
Pack of 250 for $432.00
Case of 4 PK for $1,513.00
Add to cart
22-045345
Accessories View Documents
Thermo Scientific™
C1000PK
Pink
Pack of 250 for $431.50
Case of 4 PK for $1,512.00
Add to cart
23-900-653 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
C1000TN
Tan
Case of 1000 for $1,065.00
Add to cart
22-045346
Accessories View Documents
Thermo Scientific™
C1000YL
Yellow
Pack of 250 for $438.00
Case of 4 PK for $1,524.72
Add to cart
22-050-264 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
C1000OR
Orange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-045344
Accessories View Documents
Thermo Scientific™
C1000WH
White This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-045350 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
C1000AQ
Aqua This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Handle small biopsies and fragile specimens with Thermo Scientific™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
HistoScreen™ Cassettes, Mesh Pore Insert