Learn More
Fisherbrand™ Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes
Fisher Scientific™ Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes are innovative, disposable and make biopsy pads obsolete.
 View more versions of this product
Catalog No. 15-182-705F
Add to cart
Description
Description
  • Molded from a special high-density polymer
  • Mesh design features more than 2000, 0.26mm “pores” for maximum fluid exchange and drainage
  • One piece, integral lid eliminates the need for separate lids
  • Lids can be opened or closed as many times as needed and relocked securely each time
  • 45° anterior writing surface suitable for use with certain types of labeling instruments
  • Six-Compartment Cassette holds up to six tissue specimens; each one placed in a separate 0.25 x 0.43 in. (7 x 12mm) numbered compartment
Specifications
Specifications
Mesh
45°
Six Compartments
Biopsy Cassette
Biopsy Cassette
Peach
High Density Polymer
SDS
Documents
Accessories