Learn More
Electron Microscopy SciencesSupplier Diversity Partner Tissue-Tek Cryo Mold Intermediate, Model 4566 DFS Item
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 6253415
These disposable plastic Cryomolds produce a uniformly shaped flat surface specimen block with tissue freezing medium (O.C.T.) for frozen sectioning. Standard Cryomold fits directly into Cryobar freezing wells of the Tissue-Tek II microtome/cryostat. Use a Cryomold adapter to adapt smaller biopsy and intermediate Cryomold molds to freezing wells on the Cryobar unit. Use Cryomold adapter for freezing wells on the Cryobar. Size: 15x15x5 mm.
Model: 4566
Catalog No. NC1466760