Learn More
Thermo Scientific™ TissueLoc™ Tissue Cassettes Available on GSA/VA Contract for Federal Government customers only.
Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Thermo Scientific™ TissueLoc™ Tissue Cassettes.
Manufacturer:  Thermo Scientific™ 58962
 View more versions of this product
Catalog No. 22-050-171
  Check Availability
Add to cart
Specifications
Description
Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Thermo Scientific™ TissueLoc™ Tissue Cassettes. These cassettes are preassembled for added convenience.
  • Three-sided writing surface ensures positive identification from grossing to filing
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Eleven colors allow for easy specimen coding
Description & Specifications
Specifications
Hinged
Aqua