Thermo Scientific™ TissueLoc™ Tissue Cassettes Available on GSA/VA Contract for Federal Government customers only.

Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Thermo Scientific™ TissueLoc™ Tissue Cassettes.

Manufacturer: Thermo Scientific™ 58963

 View more versions of this product

Catalog No. 22-050-172

  Check Availability
Add to Cart

    Specifications

    Closure Type Hinged
    Color Gold

    Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Thermo Scientific™ TissueLoc™ Tissue Cassettes. These cassettes are preassembled for added convenience.

    • Three-sided writing surface ensures positive identification from grossing to filing
    • Solvent- and decalcifier-resistant and compatible with microwave techniques
    • Eleven colors allow for easy specimen coding

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit