Learn More
Thermo Scientific™ TissueLoc™ Tissue Cassettes IV in Tube Packs Available on GSA/VA Contract for Federal Government customers only.
Aid the process of speedy and reliable specimen labeling with Thermo Scientific™ TissueLoc™ Tissue Cassettes IV in Tube Packs.
Manufacturer:  Thermo Scientific™ 41723T
 View more versions of this product
Catalog No. 41723T
  Check Availability
Add to cart
Specifications
Description
Aid the process of speedy and reliable specimen labeling with Thermo Scientific™ TissueLoc™ Tissue Cassettes IV in Tube Packs, compatible with Thermo Scientific™ MicroWriter™ and PrintMate™ cassette labelers.
  • Designed with slotted-style pores
  • Packaged in 10 tubes of 75 cassettes each
Description & Specifications
Specifications
Slotted
Green