Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TissueLoc™ Tissue Cassettes

Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Thermo Scientific™ TissueLoc™ Tissue Cassettes.

Manufacturer:  Thermo Scientific™ 58958

 View more versions of this product

Catalog No. 22-050-167


  Check Availability
Add to cart

Specifications

Description

Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Thermo Scientific™ TissueLoc™ Tissue Cassettes. These cassettes are preassembled for added convenience.

  • Three-sided writing surface ensures positive identification from grossing to filing
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Eleven colors allow for easy specimen coding
Description & Specifications

Specifications

Tissue Cassettes, Slotted Pore
Hinged
Pink
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit