Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Titanium Buckets for SureSpin™ 632 rotor
For use with SureSpin™ 632 rotor
$19150.00
Specifications
For Use With (Equipment) | SureSpin 632 |
---|---|
Material | Titanium |
Quantity | 6/Pk. |
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
75-003-036
|
Thermo Scientific™
75003036 |
17 mL |
Pack of 6 for $19,150.00
|
|
75-003-037
|
Thermo Scientific™
75003037 |
38.5 mL |
Pack of 6 for $19,150.00
|
|
Specifications
SureSpin 632 | |
Titanium | |
6/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title