Titanium carbide, 99.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 40178A1

Quantity 1kg

 View more versions of this product

Catalog No. AA40178A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12070-08-5
    Physical Form Typically 2 Micron Powder
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula CTi
    Formula Weight 59.91
    MDL Number MFCD00011268
    Chemical Name or Material Titanium carbide

    Specifications

    Formula Weight 59.91
    Quantity 1kg
    EINECS Number 235-120-4
    Physical Form Typically 2 Micron Powder
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula CTi
    MDL Number MFCD00011268
    CAS 12070-08-5
    Chemical Name or Material Titanium carbide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit