Learn More
Titanium carbide, nanopowder, Thermo Scientific Chemicals
Description
Titanium carbide is used in the manufacture of wear-resistant tools, cutting tools, As a coating for abrasive steel bearings, In the form of nano titanium carbide ceramic in optics applications, Enhance the conductivity of materials and as a nucleating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 4820°C |
Melting Point | ∽3140°C |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Soluble in nitric acid and aqua regia. Insoluble in water. |
Formula Weight | 59.91 |
Density | 4.93 |
Chemical Name or Material | Titanium carbide |
Safety and Handling
Hazard Category | H228 |
Precautionary Statement | P210-P240-P241-P280-P370+P378q |
DOTInformation | Hazard Class: 4.1; Packaging Group: III |
EINECSNumber | 235-120-4 |
RTECSNumber | XR1903500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.