Alfa Aesar™ Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol

Manufacturer: Alfa Aesar™ 8936022

Quantity 100g

 View more versions of this product

Catalog No. AA8936022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17927-72-9
    Molecular Formula C16H28O6Ti
    Formula Weight 364.3
    MDL Number MFCD00000031
    UN Number UN1219
    Synonym Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)
    Chemical Name or Material Titanium (diisopropoxide) bis (2,4-pentanedionate)

    Specifications

    Formula Weight 364.3
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Sensitivity Moisture Sensitive
    MDL Number MFCD00000031
    CAS 17927-72-9
    Flash Point 18°C (61°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    TSCA TSCA
    Odor Alcohol-like
    Molecular Formula C16H28O6Ti
    UN Number UN1219
    Color Brown
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Refractive Index 1.494
    Synonym Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)
    Quantity 100g
    Concentration 75% in isopropanol
    Density 1.010g/mL
    Chemical Name or Material Titanium (diisopropoxide) bis (2,4-pentanedionate)

    GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit