Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Foil, 0.025mm (0.001 in.) thick, 99.94% (Metals basis), Alfa Aesar™

Click to view available options
Quantity:
25 x 75mm
75 x 75mm
150 x 150mm
7440-32-6
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
7440-32-6
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]

Form: Approx. 0.2g/25 x 75mm

Quantity 150 x 150mm
Appearance Silver Gray foil
Assay Percent Range 99.94% (Metals basis)
Odor Odorless
Physical Form Foil, 0.025mm (0.001 in.) thick
Color Silver to Gray
Chemical Name or Material Titanium

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures