Titanium Foil, 0.89mm (0.035 in.) thick, 99.7% (Metals basis), Alfa Aesar™

Form: Approx. 40g/100 x 100mm

Manufacturer: Alfa Aesar 10399GW

Quantity 200 x 200mm

 View more versions of this product

Catalog No. AA10399GW

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-32-6
    Physical Form Foil, 0.89mm (0.035 in.) thick
    Assay Percent Range 99.7% (Metals basis)
    MDL Number MFCD00011264
    Chemical Name or Material Titanium

    Specifications

    Quantity 200 x 200mm
    Color Dark gray
    Physical Form Foil, 0.89mm (0.035 in.) thick
    Assay Percent Range 99.7% (Metals basis)
    MDL Number MFCD00011264
    CAS 7440-32-6
    Odor Odorless
    Appearance Dark Gray foil
    Chemical Name or Material Titanium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit