Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium foil, 99% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
50 x 50mm
150 x 150mm
7440-32-6
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
7440-32-6
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
Quantity 150 x 150mm
Assay Percent Range 99% (metals basis)
Physical Form ≈1.4g/50x50mm
Chemical Name or Material Titanium foil

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures