Titanium(II) Oxide, 99.5% (Metal Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 7712606

Quantity 5g

 View more versions of this product

Catalog No. AA7712606

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range 99.5%, metal basis
    Molecular Formula TiO
    Formula Weight 63.9
    MDL Number MFCD00036279
    Chemical Name or Material Titanium(II) Oxide

    Specifications

    Formula Weight 63.9
    Quantity 5g
    Assay Percent Range 99.5%, metal basis
    Molecular Formula TiO
    MDL Number MFCD00036279
    CAS 12137-20-1
    Chemical Name or Material Titanium(II) Oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit