Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(III) fluoride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
13470-08-1
F3Ti
104.9
MFCD00042544
13470-08-1
F3Ti
104.9
MFCD00042544
Reduction reagent
Assay Percent Range 97.5% min. (ex F)
Linear Formula TiF3
Solubility Information Solubility in water: hydrolyses
Formula Weight 104.9
Physical Form Crystalline Powder
Percent Purity 98%
Identification Confirmed
Packaging Plastic Bottle
Boiling Point 1400°C
Color Brown to Red
Melting Point 1200°C
Quantity 5g
Chemical Name or Material Titanium(III) fluoride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-732-4