Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) bromide, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
7789-68-6
TiBr4
367.5
MFCD00011265
7789-68-6
TiBr4
367.5
MFCD00011265
Assay Percent Range 98%
Linear Formula TiBr4
Merck Index 15,9633
Solubility Information Solubility in water: reacts. Other solubilities: soluble in absolute ether and absolute alcohol
Formula Weight 367.5
Percent Purity 98%
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in absolute ether and absolute alcohol
Boiling Point 230.0°C
Melting Point 39.0°C
Quantity 5g
Chemical Name or Material Titanium(IV) bromide
Show More Show Less

EINECSNumber : 232-185-0