Titanium(IV) bromide, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 3959209

Quantity 10g

 View more versions of this product

Catalog No. AA3959209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-68-6
    Physical Form Crystalline
    Assay Percent Range 98%
    Molecular Formula TiBr4
    Formula Weight 367.54
    MDL Number MFCD00011265

    Specifications

    Formula Weight 367.54
    Quantity 10g
    Color Orange
    Physical Form Crystalline
    Odor Acrid
    Assay Percent Range 98%
    Molecular Formula TiBr4
    MDL Number MFCD00011265
    CAS 7789-68-6

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit