Learn More
Alfa Aesar™ Titanium(IV) butoxy isopropoxy complexes, VERTEC™ BIP
Manufacturer:  Alfa Aesar™ 4468122
Quantity 100g
 View more versions of this product
Catalog No. AA4468122
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
68955-22-6
MFCD01324803
UN1993
Titanium (IV) butoxy isopropoxy complexes, VERTEC™ BIP
Specifications
68955-22-6
MFCD01324803
100g
Liquid
UN1993
Titanium (IV) butoxy isopropoxy complexes, VERTEC™ BIP
Safety and Handling
GHS H Statement
H318-H226-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.