Alfa Aesar™ Titanium(IV) chloride, 1M soln. in toluene

Manufacturer: Alfa Aesar™ H31808AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH31808AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7550-45-0
    Molecular Formula Cl4Ti
    Linear Formula TiCl4
    Formula Weight 189.69
    MDL Number MFCD00011267
    UN Number UN2924
    Synonym Titanium tetrachloride; Titanic chloride
    Chemical Name or Material Titanium(IV) chloride

    Specifications

    Formula Weight 189.69
    Linear Formula TiCl4
    MDL Number MFCD00011267
    UN Number UN2924
    Synonym Titanium tetrachloride; Titanic chloride
    Flash Point 8°C (46°F)
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 1M soln. in toluene
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    TSCA TSCA
    Molecular Formula Cl4Ti
    CAS 7550-45-0
    Quantity 100mL
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H224-H304-H361-H373-H314-H318-H336
    Extremely flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    Chemical Name or Material Titanium(IV) chloride

    Description

    Applications
    Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

    Solubility
    Reacts with water.

    Notes
    Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

    GHS H Statement
    H224-H304-H361-H373-H314-H318-H336
    Extremely flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit