Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) chloride, 1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 378168000

 View more versions of this product

Catalog No. AC378168000


Order in Bulk

Request bulk or custom formats

Due to product restrictions, please to purchase or view availability for this product.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Titanium(IV) chloride
Orange
Liquid
Cl4Ti
8°C
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
XJDNKRIXUMDJCW-UHFFFAOYSA-J
titanium(4+);tetrachloride
160960
0.95 to 1.1M
0.964
1M Solution in Toluene
7550-45-0, 108-88-3
800 mL
MFCD00011267
15, 9634
Solubility in water: reacts
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
189.71
189.71
AcroSeal™ Glass Bottle
0.9640g/mL
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Suspected of damaging the unborn child.
May cause drowsiness or dizziness

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for seve

GHS Signal Word: Danger

EINECSNumber : 231-441-9

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

RUO – Research Use Only