Learn More
Titanium(IV) chloride, 1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Specifications
Chemical Name or Material | Titanium(IV) chloride |
---|---|
Name Note | 1M Solution in Toluene |
CAS | 7550-45-0, 108-88-3 |
Color | Orange |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC378161000
|
Thermo Scientific Chemicals
378161000 |
100 mL | AcroSeal™ Glass bottle |
Each
for $50.65
Save 20% Reg :
Order in Bulk
Request bulk or custom formats |
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
AC378168000
|
Thermo Scientific Chemicals
378168000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $268.00
Order in Bulk
Request bulk or custom formats |
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Titanium(IV) chloride | |
7550-45-0, 108-88-3 | |
Liquid | |
MFCD00011267 | |
15, 9634 | |
Solubility in water: reacts | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] | |
189.71 | |
189.71 | |
0.964 |
1M Solution in Toluene | |
Orange | |
Cl4Ti | |
8°C | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
titanium(4+);tetrachloride | |
160960 | |
0.95 to 1.1M | |
0.9640g/mL |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Suspected of damaging the unborn child.
May cause drowsiness or dizziness
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for seve
GHS Signal Word: Danger
EINECSNumber : 231-441-9
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.