Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) chloride, 99.0% min, typically 99.6%, Alfa Aesar™

For Use in Manufacture of Titanium Pigments and Metal, As a Catalyst in organic Syntheses, in Iridescent Glass

Manufacturer:  Alfa Aesar 14713L7

Quantity 3 x 1L

 View more versions of this product

Catalog No. AA14713L7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7550-45-0
99.0% min., typically 99.6%
189.69
Titanic chloride
Liquid
Cl4Ti
MFCD00011267
Titanium(IV) chloride

Specifications

7550-45-0
99.0% min., typically 99.6%
Colorless to yellow liquid
MFCD00011267
1.726g/cm3
3 x 1L
Liquid
Cl4Ti
189.69
Titanic chloride
Clear yellow
Titanium(IV) chloride

Safety and Handling

SDS & Safety Documents

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 6.1, Packing Group: I

EINECSNumber : 231-441-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit