Titanium(IV) chloride, 99.0% min, typically 99.6%, Alfa Aesar™

For Use in Manufacture of Titanium Pigments and Metal, As a Catalyst in organic Syntheses, in Iridescent Glass

Manufacturer: Alfa Aesar 14713AE

Quantity 100mL

 View more versions of this product

Catalog No. AA14713AE

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7550-45-0
    Physical Form Liquid
    Assay Percent Range 99.0% min., typically 99.6%
    Molecular Formula Cl4Ti
    Formula Weight 189.69
    MDL Number MFCD00011267
    Synonym Titanic chloride
    Chemical Name or Material Titanium(IV) chloride

    Specifications

    Color Clear yellow
    Physical Form Liquid
    Formula Weight 189.69
    Quantity 100mL
    DOT Information Hazard Class: 6.1, Packing Group: I
    Density 1.726g/cm3
    EINECS Number 231-441-9
    Assay Percent Range 99.0% min., typically 99.6%
    Molecular Formula Cl4Ti
    Appearance Colorless to yellow liquid
    MDL Number MFCD00011267
    CAS 7550-45-0
    Synonym Titanic chloride
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    Chemical Name or Material Titanium(IV) chloride

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit