Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Titanium(IV) ethoxide, TiO2 33% min

Manufacturer:  Alfa Aesar™ 7714218

Quantity 50g

 View more versions of this product

Catalog No. AA7714218


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3087-36-3
Ti(OCH2CH3)4
MFCD00009071
Tetraethyl titanate
C8H20O4Ti
228.15
UN2924
Titanium (IV) ethoxide

Specifications

3087-36-3
C8H20O4Ti
228.15
UN2924
Soluble in organic solvents; Decomposes in water
1.088g/mL
1.5043
150° to 152°C (10mmHg)
−40°C
28°C (84°F)
Liquid
Ti(OCH2CH3)4
MFCD00009071
Tetraethyl titanate
TiO2 33% min
Alcohol-like
Moisture Sensitive
Pale yellow
50g
Titanium (IV) ethoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit