Alfa Aesar™ Titanium(IV) ethoxide, VERTEC™ ET, 99+%

Manufacturer: Alfa Aesar™ 4467030

Quantity 250g

 View more versions of this product

Catalog No. AA4467030

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 3087-36-3
    Molecular Formula C8H20O4Ti
    Linear Formula Ti(OCH2CH3)4
    Formula Weight 228.15
    MDL Number MFCD00009071
    UN Number UN3181
    Synonym Tetraethyl orthotitanate; Tetraethyl titanate
    Chemical Name or Material Titanium (IV) ethoxide, VERTEC™ ET

    Specifications

    Physical Form Low Melting Solid
    Formula Weight 228.15
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/.../equipment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Melting Point <60°C
    Molecular Formula C8H20O4Ti
    UN Number UN3181
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Odor Alcohol-like
    Health Hazard 1 Warning
    TSCA TSCA
    MDL Number MFCD00009071
    CAS 3087-36-3
    Color Orange
    Quantity 250g
    Flash Point 33°C (91°F)
    Refractive Index 1.5043
    Synonym Tetraethyl orthotitanate; Tetraethyl titanate
    Health Hazard 2 GHS H Statement
    H228-H319
    Flammable solid.
    Causes serious eye irritation.
    Linear Formula Ti(OCH2CH3)4
    Boiling Point 122°C (1mmHg)
    Chemical Name or Material Titanium (IV) ethoxide, VERTEC™ ET
    Assay ≥99%

    GHS H Statement
    H228-H319
    Flammable solid.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P240-P241-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/.../equipment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit