Alfa Aesar™ Titanium(IV) isopropoxide, 97+%

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids

Manufacturer: Alfa Aesar™ 7711509

Quantity 10g

 View more versions of this product

Catalog No. AA7711509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 546-68-9
    Molecular Formula C12H28O4Ti
    Linear Formula Ti[OCH(CH3)2]4
    Formula Weight 284.23
    MDL Number MFCD00008871
    UN Number UN2413
    Beilstein 3679474
    Merck Index 14,9480
    Synonym Tetraisopropoxytitanium(IV); Tetraisopropyl orthotitanate
    Chemical Name or Material Titanium (IV) isopropoxide

    Specifications

    Formula Weight 284.23
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00008871
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
    Physical Form Liquid
    Refractive Index 1.4640
    Beilstein 3679474
    Quantity 10g
    Flash Point 46°C (115°F)
    Health Hazard 1 Warning
    RTECS Number NT8060000
    Sensitivity Moisture Sensitive
    CAS 546-68-9
    UN Number UN2413
    Merck Index 14,9480
    Color Colorless to pale yellow
    TSCA TSCA
    Molecular Formula C12H28O4Ti
    Linear Formula Ti[OCH(CH3)2]4
    Synonym Tetraisopropoxytitanium(IV); Tetraisopropyl orthotitanate
    Boiling Point 232°C
    Solubility Information Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water
    Health Hazard 2 GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.
    Density 0.955g/mL
    Melting Point 16° to 20°C
    Chemical Name or Material Titanium (IV) isopropoxide
    Assay ≥97%

    GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit