Alfa Aesar™ Titanium(IV) isopropoxide, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 3579422

Quantity 100g

 View more versions of this product

Catalog No. AA3579422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 546-68-9
    Linear Formula Ti[OCH(CH3)2]4
    Formula Weight 284.23
    MDL Number MFCD00008871
    UN Number UN2413
    Beilstein 3679474
    Merck Index 14,9480
    Synonym Tetraisopropoxytitanium(IV); Tetraisopropyl orthotitanate
    Chemical Name or Material Titanium(IV) isopropoxide

    Specifications

    Formula Weight 284.23
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.
    UN Number UN2413
    Refractive Index 1.4640
    Color Colorless
    Quantity 100g
    MDL Number MFCD00008871
    Beilstein 3679474
    Merck Index 14,9480
    Sensitivity Moisture Sensitive
    Flash Point 46° (115°F)
    TSCA TSCA
    Health Hazard 1 Warning
    Density 0.955g/mL
    Melting Point 16° to 20°C
    Boiling Point 232°C
    Linear Formula Ti[OCH(CH3)2]4
    CAS 546-68-9
    Synonym Tetraisopropoxytitanium(IV); Tetraisopropyl orthotitanate
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
    RTECS Number NT8060000
    Chemical Name or Material Titanium(IV) isopropoxide
    Assay 99.995%, (metals basis)

    GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit