Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Titanium(IV) methoxide, 95%

Manufacturer:  Alfa Aesar™ 7713736

Quantity 500g

 View more versions of this product

Catalog No. AA7713736


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

992-92-7
MFCD00014883
Methyl orthotitanate; Methyl titanate(IV)
Ti(OCH3)4·(CH3OH)x, x<1
UN3181
Titanium (IV) methoxide

Specifications

992-92-7
Ti(OCH3)4·(CH3OH)x, x<1
UN3181
Moisture Sensitive
200° to 210°C
48°C (118°F)
95%
Granular/Lump
MFCD00014883
Methyl orthotitanate; Methyl titanate(IV)
>243°C (52mmHg)
500g
Titanium (IV) methoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H302-H312-H332-H319
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P304+P340-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit