Alfa Aesar™ Titanium(IV) methoxide, 95%

Manufacturer: Alfa Aesar™ 7713706

Quantity 5g

 View more versions of this product

Catalog No. AA7713706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 992-92-7
    Molecular Formula Ti(OCH3)4·(CH3OH)x, x<1
    MDL Number MFCD00014883
    UN Number UN3181
    Synonym Methyl orthotitanate; Methyl titanate(IV)
    Chemical Name or Material Titanium (IV) methoxide

    Specifications

    Physical Form Granular/Lump
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P304+P340-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point >243°C (52mmHg)
    Sensitivity Moisture Sensitive
    MDL Number MFCD00014883
    Synonym Methyl orthotitanate; Methyl titanate(IV)
    Melting Point 200° to 210°C
    Flash Point 48°C (118°F)
    CAS 992-92-7
    UN Number UN3181
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H302-H312-H332-H319
    Flammable solid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    TSCA TSCA
    Molecular Formula Ti(OCH3)4·(CH3OH)x, x<1
    Quantity 5g
    Chemical Name or Material Titanium (IV) methoxide
    Assay 95%

    GHS H Statement
    H228-H302-H312-H332-H319
    Flammable solid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P304+P340-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit