Alfa Aesar™ Titanium(IV) methylphenoxide, tech.

Manufacturer: Alfa Aesar™ 8945430

Quantity 250g

 View more versions of this product

Catalog No. AA8945430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 28503-70-0
    Molecular Formula C28H28O4Ti
    Formula Weight 476.43
    MDL Number MFCD00061534
    Synonym Titanium(IV) cresylate
    Chemical Name or Material Titanium (IV) methylphenoxide
    Grade Technical Grade

    Specifications

    Formula Weight 476.43
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point −36°C
    MDL Number MFCD00061534
    CAS 28503-70-0
    Synonym Titanium(IV) cresylate
    Color Dark red
    Density 1.055g/mL
    TSCA TSCA
    Physical Form Liquid
    Health Hazard 1 Warning
    Sensitivity Moisture Sensitive
    Molecular Formula C28H28O4Ti
    Boiling Point 230° to 240°C (0.01mmHg)
    Quantity 250g
    Flash Point 66°C (150°F)
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Chemical Name or Material Titanium (IV) methylphenoxide
    Grade Technical Grade

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit