Alfa Aesar™ Titanium(IV) n-propoxide

Manufacturer: Alfa Aesar™ 7714114

Quantity 25g

 View more versions of this product

Catalog No. AA7714114

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 3087-37-4
    Molecular Formula Ti(OCH2CH2CH3)4
    Formula Weight 284.25
    MDL Number MFCD00009365
    UN Number UN2413
    Synonym Tetrapropyl orthotitanate(IV)
    Chemical Name or Material Titanium (IV) n-propoxide

    Specifications

    Formula Weight 284.25
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    UN Number UN2413
    Boiling Point >58°C (1mmHg)
    Quantity 25g
    Health Hazard 1 Warning
    Density 0.9550g/mL
    Molecular Formula Ti(OCH2CH2CH3)4
    MDL Number MFCD00009365
    Synonym Tetrapropyl orthotitanate(IV)
    Melting Point >20°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
    TSCA TSCA
    Odor Alcohol-like
    Refractive Index 1.4986
    CAS 3087-37-4
    Flash Point 42^o (109^oF)
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Titanium (IV) n-propoxide

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit