Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxide, 99.999%, (trace metal basis), predominantly rutile, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13463-67-7
O2Ti
79.87
MFCD00011269,MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O
13463-67-7
O2Ti
79.87
MFCD00011269,MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula TiO2
Merck Index 15,9628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
Formula Weight 79.88
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Density 4.2300g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
Specific Gravity 4.23
Total Trace Metal Impurities 0.001% max
Boiling Point 2900.0°C
Color White
Melting Point 1855.0°C
Quantity 5g
Chemical Name or Material Titanium(IV) oxide, 99.999%
Show More Show Less

EINECSNumber : 236-675-5

RTECSNumber : XR2275000

TSCA : TSCA