Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxide, anatase, 99.6% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3620014

Quantity 25g

 View more versions of this product

Catalog No. AA3620014

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1317-70-0
79.9
14,9472
TiO2
MFCD00011269
Titanium(IV) oxide, anatase

Specifications

1317-70-0
TiO2
MFCD00011269
3.9
25g
99.6%, (metals basis)
3 to 6mm Sintered Pieces
79.9
14,9472
Odorless
Titanium(IV) oxide, anatase

Safety and Handling

GHS H Statement
H351;
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a;
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BV6550000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit