Titanium(IV) oxide, anatase, 99.6% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 36200-22

Quantity 100g

 View more versions of this product

Catalog No. AA3620022

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1317-70-0
    Molecular Formula TiO2
    Formula Weight 79.9
    MDL Number MFCD00011269
    Merck Index 14,9472
    Chemical Name or Material Titanium(IV) oxide, anatase

    Specifications

    Formula Weight 79.9
    Quantity 100g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351;
    Suspected of causing cancer.
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a;
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Odorless
    Physical Form 3 to 6mm Sintered Pieces
    Molecular Formula TiO2
    MDL Number MFCD00011269
    CAS 1317-70-0
    Merck Index 14,9472
    Density 3.9
    RTECS Number BV6550000
    Chemical Name or Material Titanium(IV) oxide, anatase
    Assay 99.6%, (metals basis)

    GHS H Statement
    H351;
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a;
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit