Titanium(IV) oxide, anatase, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3995336

Quantity 500g

 View more versions of this product

Catalog No. AA3995336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-70-0
    Physical Form 32Nm APS Powder, S.A. 45m&superscript_2;/g
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula TiO2
    Formula Weight 79.9
    MDL Number MFCD00011269
    Chemical Name or Material Titanium(IV) oxide

    Specifications

    Physical Form 32Nm APS Powder, S.A. 45m&superscript_2;/g
    Formula Weight 79.9
    Quantity 500g
    EINECS Number 215-280-1
    Density 3.9g/mL
    Molecular Formula TiO2
    MDL Number MFCD00011269
    CAS 1317-70-0
    Assay Percent Range 99.9% (metals basis)
    Chemical Name or Material Titanium(IV) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit