Alfa Aesar™ Titanium(IV) oxide, anatase, nanopowder, 99% (metals basis)

Manufacturer: Alfa Aesar™ 4721514

Quantity 25g

 View more versions of this product

Catalog No. AA4721514

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1317-70-0
    Formula Weight 79.9
    MDL Number MFCD00011269
    Merck Index 14,9472
    Chemical Name or Material Titanium(IV) oxide, Anatase

    Specifications

    Formula Weight 79.9
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Color White to off-white
    Density 3.9g/mL
    Particle Size 10 to 30nm APS
    Surface Area 200 to 220m2/g
    MDL Number MFCD00011269
    CAS 1317-70-0
    Merck Index 14,9472
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Nanopowder
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    RTECS Number BV6550000
    Chemical Name or Material Titanium(IV) oxide, Anatase
    Assay 99%, (metals basis)

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit