Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Titanium(IV) oxide, naturally occurring mineral, grains, approximately 0.06-0.19 in.

For grain mounts, XRD and microprobe standards, identification of unknown minerals

Manufacturer:  Alfa Aesar™ 4264806

Quantity 5g

 View more versions of this product

Catalog No. AA4264806


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13463-67-7
79.9
MFCD00011269
Titanium (IV) oxide, Naturally occurring mineral, grains, approximately 0.06 to 0.19 in.

Specifications

13463-67-7
79.9
4.26g/mL
2.61
White
5g
Grains
MFCD00011269
Odorless
2500° to 3000°C
1830° to 1850°C
Titanium (IV) oxide, Naturally occurring mineral, grains, approximately 0.06 to 0.19 in.

Safety and Handling

SDS & Safety Documents

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XR2275000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit