Alfa Aesar™ Titanium(IV) oxide, naturally occurring mineral, grains, approximately 0.06-0.19 in.

For grain mounts, XRD and microprobe standards, identification of unknown minerals

Manufacturer: Alfa Aesar™ 4264803

Quantity 1g

 View more versions of this product

Catalog No. AA4264803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13463-67-7
    Formula Weight 79.9
    MDL Number MFCD00011269
    Chemical Name or Material Titanium (IV) oxide, Naturally occurring mineral, grains, approximately 0.06 to 0.19 in.

    Specifications

    Formula Weight 79.9
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Grains
    Color White
    Quantity 1g
    Density 4.26g/mL
    RTECS Number XR2275000
    TSCA TSCA
    Refractive Index 2.61
    CAS 13463-67-7
    Boiling Point 2500° to 3000°C
    Melting Point 1830° to 1850°C
    Health Hazard 1 Warning
    MDL Number MFCD00011269
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    Chemical Name or Material Titanium (IV) oxide, Naturally occurring mineral, grains, approximately 0.06 to 0.19 in.

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit