Learn More
Titanium(IV) oxide, rutile, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Titanium(IV) oxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2500°C to 3000°C |
Melting Point | 1830°C to 1850°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Merck Index | 14,9472 |
Solubility Information | Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water. |
Formula Weight | 79.9 |
Percent Purity | 99.99% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H351 |
Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 215-282-2 |
RTECSNumber | XR2275000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.