Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) tert-butoxide, 98%, Thermo Scientific Chemicals

Catalog Number p-200653
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s, px6yr1s99s, titanium iv tert-butoxide, titanium tetra-tert-butoxide, titanium tert-butoxide, titanum tetra t-butoxide, tetra-tert-butoxytitanium iv, titanium 4+ 2-methylpropan-2-olate
6451515
[Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s, px6yr1s99s, titanium iv tert-butoxide, titanium tetra-tert-butoxide, titanium tert-butoxide, titanum tetra t-butoxide, tetra-tert-butoxytitanium iv, titanium 4+ 2-methylpropan-2-olate
6451515
[Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8893g/mL
Boiling Point 160.0°C to 165.0°C (15.0 mmHg)
Flash Point 47°C
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula Ti[OC(CH3)3]4
Quantity 5 g
Beilstein 01, IV, 1613
Specific Gravity 0.8893
IUPAC Name titanium(4+) tetrakis(2-methylpropan-2-olate)
Formula Weight 340.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Titanium(IV) tert-butoxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 221-412-9

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit