Learn More
Alfa Aesar™ Titanium Nickel powder composite, -325 mesh
Manufacturer:  Alfa Aesar™ 8838436
Quantity 500g
 View more versions of this product
Catalog No. AA8838436
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD02091734
UN3089
Titanium Nickel powder composite
Specifications
Ti:Ni; 70:30 wt%
Powder
UN3089
Air Sensitive
500g
MFCD02091734
−325 Mesh
Titanium Nickel powder composite
Safety and Handling
SDS & Safety Documents
GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
TSCA : TSCA